missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-fluorobenzaldehyde 98.0+%, TCI America™

Catalog No. p-7124055
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B53635G 5 g
B536325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B53635G Supplier TCI America Supplier No. B53635G

Chemical Identifiers

CAS 149947-15-9
Molecular Formula C7H4BrFO
Molecular Weight (g/mol) 203.01
MDL Number MFCD09027089
InChI Key OUAZPCKRSSEQKB-UHFFFAOYSA-N
Synonym 3-bromo-2-fluorobenzenecarbaldehyde, 3-bromo-2-fluorobenzadehyde, 2-fluoro-3-bromobenzaldehyde, benzaldehyde,3-bromo-2-fluoro, benzaldehyde, 3-bromo-2-fluoro, 3-bromofluorobenzaldehyde, intermediates-zcf02145, acmc-209d3i, 3-bromo-2-fluoro-benzaldehyde, 3-bromanyl-2-fluoranyl-benzaldehyde
PubChem CID 22019361
IUPAC Name 3-bromo-2-fluorobenzaldehyde
SMILES C1=CC(=C(C(=C1)Br)F)C=O

Specifications

Melting Point 43°C
Color White-Yellow
Quantity 5 g
Formula Weight 203.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-2-fluorobenzaldehyde