missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-fluoroacetophenone, 96%, Thermo Scientific Chemicals

Catalog No. AAH62708MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62708MD 250 mg
AAH6270803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH62708MD Supplier Thermo Scientific Chemicals Supplier No. H62708MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 161957-61-5 | Molecular Formula: C8H6BrFO | Molecular Weight: 217.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 161957-61-5
Molecular Formula C8H6BrFO
Molecular Weight (g/mol) 217.04
MDL Number MFCD09264507
InChI Key GSLFYQYBXIVNLS-UHFFFAOYSA-N
Synonym 3'-bromo-2'-fluoroacetophenone, 1-3-bromo-2-fluorophenyl ethanone, 1-3-bromo-2-fluorophenyl ethan-1-one, 1-3-bromo-2-fluoro-phenyl-ethanone, 3-bromo-2-fluoroacetophenone, 1-3-bromo-2-fluoro-phenyl ethanone, 2'-fluoro-3'-bromo acetophenone, ethanone, 1-3-bromo-2-fluorophenyl, 1-acetyl-3-bromo-2-fluorobenzene, pubchem4095
PubChem CID 14937359
IUPAC Name 1-(3-bromo-2-fluorophenyl)ethanone
SMILES CC(=O)C1=C(F)C(Br)=CC=C1

Specifications

Quantity 250 mg
Formula Weight 217.04
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Bromo-2-fluoroacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only