accessibility menu, dialog, popup

UserName

H3COBrF

CAS RN 161957-61-5

IUPAC Name: 1-(3-bromo-2-fluorophenyl)ethan-1-one
Synonyms: 1-(3-bromo-2-fluorophenyl)ethanone
Molecular Weight (g/mol): 217.04
Molecular Formula: C8H6BrFO
InChi Key: GSLFYQYBXIVNLS-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(F)C(Br)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results