missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-2-ethoxypyridine, 95%, Thermo Scientific Chemicals
Description
3-Bromo-2-ethoxypyridin is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Bromo-2-ethoxypyridin is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
3-Bromo-2-ethoxypyridin is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
Specifications
Specifications
| Quantity | 1 g |
| Synonym | 2-ethoxy-3-bromopyridine, pyridine, 3-bromo-2-ethoxy, 3-bromanyl-2-ethoxy-pyridine, 3-bromo-2-ethoxy-pyridine, pubchem6233, bromo-2 ethoxypyridine, 3-bromo-2-ethoxypyridine, pyridine,3-bromo-2-ethoxy |
| Formula Weight | 202.05 |
| Percent Purity | 95% |
| Chemical Name or Material | 3-Bromo-2-ethoxypyridine |
Safety and Handling
| Hazard Category | H302-H315-H319-H335 |
| Hazard Statement | GHS H Statement H301-H315-H319-H335 Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only