CAS RN 57883-25-7
IUPAC Name:
3-bromo-2-ethoxypyridine
Synonyms:
3-bromo-2-ethoxypyridine
Molecular Weight (g/mol):
202.05
Molecular Formula:
C7H8BrNO
InChi Key:
KLQBFCAXXZMOLZ-UHFFFAOYSA-N
SMILES:
CCOC1=C(Br)C=CC=N1
1
–
2
of
2
results
1
Promotions Available
3-Bromo-2-ethoxypyridine, 95%, Thermo Scientific Chemicals
3-Bromo-2-ethoxypyridine, 95%, Thermo Scientific Chemicals
CAS: 57883-25-7 Molecular Formula: C7H8BrNO Molecular Weight (g/mol): 202.051 MDL Number: MFCD00234309 InChI Key: KLQBFCAXXZMOLZ-UHFFFAOYSA-N PubChem CID: 10987308 IUPAC Name: 3-bromo-2-ethoxypyridine SMILES: CCOC1=C(C=CC=N1)Br