missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-chloro-5-nitropyridine 98.0+%, TCI America™

Catalog No. B40771G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40771G 1 g
B40775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40771G Supplier TCI America Supplier No. B40771G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5470-17-7
Molecular Formula C5H2BrClN2O2
Molecular Weight (g/mol) 237.437
MDL Number MFCD00233989
InChI Key PTTQIUHVDDBART-UHFFFAOYSA-N
PubChem CID 230889
IUPAC Name 3-bromo-2-chloro-5-nitropyridine
SMILES C1=C(C=NC(=C1Br)Cl)[N+](=O)[O-]

Specifications

Melting Point 68°C
Quantity 1 g
Formula Weight 237.44
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-2-chloro-5-nitropyridine