missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-(bromomethyl)propionic Acid 98.0+%, TCI America™

Catalog No. p-7119936
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
B30815G 5 g
B308125G 25 g
2 options

Catalog No. B30815G

Supplier: TCI America B30815G

Only null left

Chemical Identifiers

CAS 41459-42-1
Molecular Formula C4H5Br2O2
Molecular Weight (g/mol) 244.89
MDL Number MFCD00010643
InChI Key QQZJWQCLWOQDQV-UHFFFAOYSA-M
Synonym 3-bromo-2-bromomethyl propionic acid, 3-bromo-2-bromomethyl propanoic acid, propanoic acid, 3-bromo-2-bromomethyl, .beta.,.beta.-dibromoisobutyric acid, 3-bromo-2-bromomethyl propionicacid, bis bromomethyl acetic acid, zlchem 161, acmc-209jkh, beta,beta'-dibromoisobutyric acid, 2-bromomethyl-3-bromopropanoic acid
PubChem CID 318915
IUPAC Name 3-bromo-2-(bromomethyl)propanoate
SMILES [O-]C(=O)C(CBr)CBr

Specifications

Melting Point 99°C
Quantity 5 g
UN Number 3261
Formula Weight 245.90
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-2-(bromomethyl)propionic Acid
TSCA No
Recommended Storage Refrigerator