missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2,6-difluoroaniline, 96%

Catalog No. AAH6260903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6260903 1 g
AAH6260906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6260903 Supplier Thermo Scientific Chemicals Supplier No. H6260903

CAS: 1262198-07-1 | C6H4BrF2N | 208.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 1262198-07-1
Molecular Formula C6H4BrF2N
Molecular Weight (g/mol) 208.01
MDL Number MFCD22627833
InChI Key HIZADWSSPWORHH-UHFFFAOYSA-N
Synonym benzenamine,3-bromo-2,6-difluoro
PubChem CID 21645692
IUPAC Name 3-bromo-2,6-difluoroaniline
SMILES NC1=C(F)C=CC(Br)=C1F

Specifications

Quantity 1 g
UN Number UN2811
Formula Weight 208
Percent Purity 96%
Chemical Name or Material 3-Bromo-2,6-difluoroaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only