missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2,4,6-trimethylaniline, 98+%

Catalog No. AAL0087318
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0087318 50 g
AAL0087309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0087318 Supplier Thermo Scientific Chemicals Supplier No. L0087318
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 82842-52-2 | C9H12BrN | 214.106 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 82842-52-2
Molecular Formula C9H12BrN
Molecular Weight (g/mol) 214.106
MDL Number MFCD00015476
InChI Key MVLMPTBHZPYDBZ-UHFFFAOYSA-N
Synonym benzenamine, 3-bromo-2,4,6-trimethyl, pubchem4363, ksc496e8h, 2,4,6-trimethyl-3-bromoaniline, 3-bromo-2,4,6-trimethyl-aniline, 3-bromo-2,4,6-trimethylbenzenamine, 3-bromo-2,4,6-trimethylphenylamine, 3-bromo-2,4,6-trimethyl-phenylamine, 3-bromo-2,4,6-trimethylaniline, 3-bromo-2,4,6-trimethylaniline, 98+%
PubChem CID 688300
IUPAC Name 3-bromo-2,4,6-trimethylaniline
SMILES CC1=CC(=C(C(=C1N)C)Br)C

Specifications

Melting Point 34°C to 38°C
Boiling Point 153°C to 155°C (17 mmHg)
Flash Point >110°C (230°F)
Quantity 50 g
Beilstein 2829829
Sensitivity Light sensitive
Formula Weight 214.11
Percent Purity ≥98%
Chemical Name or Material 3-Bromo-2,4,6-trimethylaniline
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only