missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-1H-pyrrolo[2,3-b]pyridine 98.0+%, TCI America™

Catalog No. B36755G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36755G 5 g
B36751G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36755G Supplier TCI America Supplier No. B36755G

Chemical Identifiers

CAS 74420-15-8
Molecular Formula C7H5BrN2
Molecular Weight (g/mol) 197.04
MDL Number MFCD02179276
InChI Key VJDGIJDCXIEXPF-UHFFFAOYSA-N
Synonym 3-bromo-7-azaindole, 3-bromo-1h-pyrrolo 2,3-b pyridine, 3-bromo-pyrrolo 2,3-b pyridine, 1h-pyrrolo 2,3-b pyridine, 3-bromo, 3-bromo-7-azaindol, 3-bromopyrrolo 2,3-b pyridine, pubchem14707, acmc-209oup, ksc494e0h
PubChem CID 5523659
IUPAC Name 3-bromo-1H-pyrrolo[2,3-b]pyridine
SMILES BrC1=CNC2=NC=CC=C12

Specifications

Melting Point 188°C
Quantity 5 g
Formula Weight 197.04
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-1H-pyrrolo[2,3-b]pyridine