accessibility menu, dialog, popup

UserName

HNBrN

CAS RN 74420-15-8

IUPAC Name: 3-bromo-1H-pyrrolo[2,3-b]pyridine
Synonyms: 3-bromo-1H-pyrrolo[2,3-b]pyridine
Molecular Weight (g/mol): 197.04
Molecular Formula: C7H5BrN2
InChi Key: VJDGIJDCXIEXPF-UHFFFAOYSA-N
SMILES: BrC1=CNC2=NC=CC=C12
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)