missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-1H-pyrazolo[3,4-d]pyrimidin-4-amine 97.0+%, TCI America™

Catalog No. B46301G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46301G 1 g
B46305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46301G Supplier TCI America Supplier No. B46301G
Only null left

Chemical Identifiers

CAS 83255-86-1
Molecular Formula C5H4BrN5
Molecular Weight (g/mol) 214.026
MDL Number MFCD12196922
InChI Key GZQVGSRUUTUJNG-UHFFFAOYSA-N
Synonym 4-Amino-3-bromo-1H-pyrazolo[3,4-d]pyrimidine
PubChem CID 340230
IUPAC Name 3-bromo-2H-pyrazolo[3,4-d]pyrimidin-4-amine
SMILES C1=NC2=NNC(=C2C(=N1)N)Br

Specifications

Melting Point 350°C
Color White-Yellow
Quantity 1 g
Formula Weight 214.03
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-1H-pyrazolo[3,4-d]pyrimidin-4-amine