missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-1-(trimethylsilyl)-1-propyne 97.0+%, TCI America™

Catalog No. B16541G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16541G 1 g
B16545G 5 g
B165425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B16541G Supplier TCI America Supplier No. B16541G

Chemical Identifiers

CAS 38002-45-8
Molecular Formula C6H11BrSi
Molecular Weight (g/mol) 191.143
MDL Number MFCD00134460
InChI Key GAPRPFRDVCCCHR-UHFFFAOYSA-N
Synonym 3-bromo-1-trimethylsilyl-1-propyne, 3-bromoprop-1-yn-1-yl trimethylsilane, 3-bromo-1-propynyl trimethylsilane, 3-trimethylsilyl propargyl bromide, 3-bromoprop-1-ynyl trimethyl silane, silane, 3-bromo-1-propynyl trimethyl, 3-bromoprop-1-yn-1-yl trimethyl silane, acmc-1cthe, ksc496i3r
PubChem CID 642589
IUPAC Name 3-bromoprop-1-ynyl(trimethyl)silane
SMILES C[Si](C)(C)C#CCBr

Specifications

Color Yellow
Quantity 1 g
Formula Weight 191.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromo-1-(trimethylsilyl)-1-propyne