missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-1,8-naphthalic anhydride, 97%, Thermo Scientific Chemicals

Catalog No. AAL1768303
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1768303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1768303 Supplier Thermo Scientific Chemicals Supplier No. L1768303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 24050-49-5 | Molecular Formula: C12H5BrO3 | Molecular Weight: 277.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24050-49-5
Molecular Formula C12H5BrO3
Molecular Weight (g/mol) 277.07
MDL Number MFCD00228162
InChI Key LYXFXCSFCWZGNZ-UHFFFAOYSA-N
Synonym 5-bromobenzo de isochromene-1,3-dione, 3-bromo-1,8-naphthalic anhydride, 5-bromo-1h,3h-benzo de isochromene-1,3-dione, pubchem24244, 5-bromo-benzo de isochromene-1,3-dione, 1h,3h-naphtho 1,8-cd pyran-1,3-dione,5-bromo, 1h,3h-naphtho 1,8-cd pyran-1,3-dione, 5-bromo, 11-bromo-3-oxatricyclo 7.3.1.0, 1 3 trideca-1 13 ,5,7,9,11-pentaene-2,4-dione, 7-bromo-3-oxatricyclo 7.3.1.0?,__ trideca-1 13 ,5,7,9,11-pentaene-2,4-dione, 11-bromo-3-oxatricyclo 7.3.1.0?, 1 3 trideca-1 13 ,5,7,9,11-pentaene-2,4-dione
PubChem CID 3907491
IUPAC Name 7-bromo-3-oxatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione
SMILES BrC1=CC2=CC=CC3=C2C(=C1)C(=O)OC3=O

Specifications

Melting Point 244°C to 246°C
Quantity 1 g
Beilstein 180001
Sensitivity Moisture sensitive
Formula Weight 277.08
Percent Purity 97%
Chemical Name or Material 3-Bromo-1,8-naphthalic anhydride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only