missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-1,1,1-trifluoroacetone, 97%

CAS: 431-35-6 | C3H2BrF3O | 190.95 g/mol

Supplier:  Thermo Scientific Chemicals 297440050

Catalog No. AC297440050


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Bromo-1, 1, 1-trifluoroacetone
431-35-6
100.0
1.8300g/mL
5°C
96% min. (GC)
C3H2BrF3O
CF3C(O)CH2Br
5 g
3-bromo-1,1,1-trifluoroacetone, 1-bromo-3,3,3-trifluoroacetone, bromotrifluoroacetone, 3-bromo-1,1,1-trifluoro-2-propanone, 3-bromo-1,1,1-trifluoropropanone, 1-bromo-3,3,3-trifluoro-2-propanone, 2-propanone, 3-bromo-1,1,1-trifluoro, 1,1,1-trifluoro-3-bromopropanone, 3-bromo-1,1,1-trifluoro-propan-2-one, bromotrifloroacetone
ONZQYZKCUHFORE-UHFFFAOYSA-N
3-bromo-1,1,1-trifluoropropan-2-one
79008
97%
97%
96.0
Colorless to Yellow
87°C
Authentic
Glass bottle
1.3750 to 1.377
MFCD00039237
1.83
Solubility in water: soluble
FC(F)(F)C(=O)CBr
190.95
190.95
Liquid

Chemical Identifiers

431-35-6
190.95
ONZQYZKCUHFORE-UHFFFAOYSA-N
79008
FC(F)(F)C(=O)CBr
C3H2BrF3O
MFCD00039237
3-bromo-1,1,1-trifluoroacetone, 1-bromo-3,3,3-trifluoroacetone, bromotrifluoroacetone, 3-bromo-1,1,1-trifluoro-2-propanone, 3-bromo-1,1,1-trifluoropropanone, 1-bromo-3,3,3-trifluoro-2-propanone, 2-propanone, 3-bromo-1,1,1-trifluoro, 1,1,1-trifluoro-3-bromopropanone, 3-bromo-1,1,1-trifluoro-propan-2-one, bromotrifloroacetone
3-bromo-1,1,1-trifluoropropan-2-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapour.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES:

GHS Signal Word: Danger

EINECSNumber : 207-071-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only