accessibility menu, dialog, popup

UserName

FFFOBr

CAS RN 431-35-6

IUPAC Name: 3-bromo-1,1,1-trifluoropropan-2-one
Synonyms: 3-bromo-1,1,1-trifluoropropan-2-one
Molecular Weight (g/mol): 190.95
Molecular Formula: C3H2BrF3O
InChi Key: ONZQYZKCUHFORE-UHFFFAOYSA-N
SMILES: FC(F)(F)C(=O)CBr
Boiling Point 
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (3)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (3)