Learn More
3-(Boc-aminomethyl)pyridine, 97%, Thermo Scientific Chemicals
$143.11
Chemical Identifiers
| CAS | 102297-41-6 |
|---|---|
| Molecular Formula | C11H16N2O2 |
| Molecular Weight (g/mol) | 208.26 |
| MDL Number | MFCD06411296 |
| InChI Key | PHQXGCNECRISKB-UHFFFAOYSA-N |
| Synonym | n-boc-3-aminomethyl pyridine, tert-butyl pyridin-3-ylmethyl carbamate, n-boc-3-aminomethylpyridine, 3-boc-aminomethyl pyridine, tert-butyl n-pyridin-3-ylmethyl carbamate, carbamic acid, 3-pyridinylmethyl-, 1,1-dimethylethyl ester, 3-n-tert-butyloxycarbonyl amino methyl pyridine, pyridin-3-ylmethyl-carbamic acid tert-butyl ester, ambkkkkk570, tert-butyl pyridin-3-yl methylcarbamate |
| PubChem CID | 13948280 |
| IUPAC Name | tert-butyl N-(pyridin-3-ylmethyl)carbamate |
| SMILES | CC(C)(C)OC(=O)NCC1=CN=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAH5818306
|
N/A | 5 g |
Each for $143.11
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 102297-41-6 | |
| 208.26 | |
| PHQXGCNECRISKB-UHFFFAOYSA-N | |
| 13948280 | |
| CC(C)(C)OC(=O)NCC1=CN=CC=C1 |
| C11H16N2O2 | |
| MFCD06411296 | |
| n-boc-3-aminomethyl pyridine, tert-butyl pyridin-3-ylmethyl carbamate, n-boc-3-aminomethylpyridine, 3-boc-aminomethyl pyridine, tert-butyl n-pyridin-3-ylmethyl carbamate, carbamic acid, 3-pyridinylmethyl-, 1,1-dimethylethyl ester, 3-n-tert-butyloxycarbonyl amino methyl pyridine, pyridin-3-ylmethyl-carbamic acid tert-butyl ester, ambkkkkk570, tert-butyl pyridin-3-yl methylcarbamate | |
| tert-butyl N-(pyridin-3-ylmethyl)carbamate |
Specifications
| 102297-41-6 | |
| C11H16N2O2 | |
| MFCD06411296 | |
| n-boc-3-aminomethyl pyridine, tert-butyl pyridin-3-ylmethyl carbamate, n-boc-3-aminomethylpyridine, 3-boc-aminomethyl pyridine, tert-butyl n-pyridin-3-ylmethyl carbamate, carbamic acid, 3-pyridinylmethyl-, 1,1-dimethylethyl ester, 3-n-tert-butyloxycarbonyl amino methyl pyridine, pyridin-3-ylmethyl-carbamic acid tert-butyl ester, ambkkkkk570, tert-butyl pyridin-3-yl methylcarbamate | |
| CC(C)(C)OC(=O)NCC1=CN=CC=C1 | |
| 208.26 | |
| 13948280 | |
| 97% |
| 130°C | |
| 1.508 | |
| UN1993 | |
| PHQXGCNECRISKB-UHFFFAOYSA-N | |
| tert-butyl N-(pyridin-3-ylmethyl)carbamate | |
| 22°C (72°F) | |
| 208.26 | |
| 3-(Boc-aminomethyl)pyridine |
Safety and Handling
GHS H Statement
H225-H302-H317
Highly flammable liquid and vapor.
Harmful if swallowed.
May cause an allergic skin reaction.
P210-P233-P240-P241-P242-P243-P261-P264b-P270-P272-P280-P301+P312-P303+P361+P353-P330-P333+P313-P363-P370+P378q-P501c
H225-H302-H317
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only