missing translation for 'onlineSavingsMsg'
Learn More

3-(Boc-aminomethyl)pyridine, 97%, Thermo Scientific Chemicals

Catalog No. AAH5818306
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5818306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5818306 Supplier Thermo Scientific Chemicals Supplier No. H5818306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 102297-41-6
Molecular Formula C11H16N2O2
Molecular Weight (g/mol) 208.26
MDL Number MFCD06411296
InChI Key PHQXGCNECRISKB-UHFFFAOYSA-N
Synonym n-boc-3-aminomethyl pyridine, tert-butyl pyridin-3-ylmethyl carbamate, n-boc-3-aminomethylpyridine, 3-boc-aminomethyl pyridine, tert-butyl n-pyridin-3-ylmethyl carbamate, carbamic acid, 3-pyridinylmethyl-, 1,1-dimethylethyl ester, 3-n-tert-butyloxycarbonyl amino methyl pyridine, pyridin-3-ylmethyl-carbamic acid tert-butyl ester, ambkkkkk570, tert-butyl pyridin-3-yl methylcarbamate
PubChem CID 13948280
IUPAC Name tert-butyl N-(pyridin-3-ylmethyl)carbamate
SMILES CC(C)(C)OC(=O)NCC1=CN=CC=C1

Specifications

Boiling Point 130°C
Refractive Index 1.508
UN Number UN1993
Quantity 5 g
Flash Point 22°C (72°F)
Formula Weight 208.26
Percent Purity 97%
Chemical Name or Material 3-(Boc-aminomethyl)pyridine
Hazard Category H225-H302-H317
Hazard Statement GHS H Statement
H225-H302-H317
Highly flammable liquid and vapor.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P270-P272-P280-P301+P312-P303+P361+P353-P330-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only