missing translation for 'onlineSavingsMsg'
Learn More

3-(Boc-aminomethyl)benzeneboronic acid, 97%

CAS: 199609-62-6 | C12H18BNO4 | 251.089 g/mol

Supplier:  Thermo Scientific Chemicals H5245303

 View more versions of this product

Catalog No. AAH5245303


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-(Boc-aminomethyl)benzeneboronic acid
188°C to 190°C
MFCD06246052
3-n-boc-aminomethyl phenylboronic acid, 3-tert-butoxycarbonyl amino methyl phenyl boronic acid, 3-n-boc-amino methyl phenylboronic acid, 3-n-boc aminomethylphenylboronic acid, 3-tert-butoxycarbonylamino methyl phenylboronic acid, 3-n-boc-aminomethyl phenyl boronic acid, 3-tert-butoxycarbonylaminomethyl phenylboronic acid, 3-2-methylpropan-2-yl oxycarbonylamino methyl phenyl boronic acid, 3-aminomethyl benzeneboronic acid, n-boc protected, 3-tert-butoxycarbonyl amino methyl phenylboronic acid
B(C1=CC(=CC=C1)CNC(=O)OC(C)(C)C)(O)O
251.089
251.09
199609-62-6
C12H18BNO4
1 g
YHAQUGOSDQZIMA-UHFFFAOYSA-N
[3-[[(2-methylpropan-2-yl)oxycarbonylamino]methyl]phenyl]boronic acid
3684686
97%

Chemical Identifiers

199609-62-6
251.089
YHAQUGOSDQZIMA-UHFFFAOYSA-N
3684686
B(C1=CC(=CC=C1)CNC(=O)OC(C)(C)C)(O)O
C12H18BNO4
MFCD06246052
3-n-boc-aminomethyl phenylboronic acid, 3-tert-butoxycarbonyl amino methyl phenyl boronic acid, 3-n-boc-amino methyl phenylboronic acid, 3-n-boc aminomethylphenylboronic acid, 3-tert-butoxycarbonylamino methyl phenylboronic acid, 3-n-boc-aminomethyl phenyl boronic acid, 3-tert-butoxycarbonylaminomethyl phenylboronic acid, 3-2-methylpropan-2-yl oxycarbonylamino methyl phenyl boronic acid, 3-aminomethyl benzeneboronic acid, n-boc protected, 3-tert-butoxycarbonyl amino methyl phenylboronic acid
[3-[[(2-methylpropan-2-yl)oxycarbonylamino]methyl]phenyl]boronic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only