missing translation for 'onlineSavingsMsg'
Learn More

3-(Boc-aminomethyl)benzeneboronic acid, 97%

Catalog No. p-7034754
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52453MD 250 mg
AAH5245303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52453MD Supplier Thermo Scientific Chemicals Supplier No. H52453MD
Only null left

CAS: 199609-62-6 | C12H18BNO4 | 251.089 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 199609-62-6
Molecular Formula C12H18BNO4
Molecular Weight (g/mol) 251.089
MDL Number MFCD06246052
InChI Key YHAQUGOSDQZIMA-UHFFFAOYSA-N
Synonym 3-n-boc-aminomethyl phenylboronic acid, 3-tert-butoxycarbonyl amino methyl phenyl boronic acid, 3-n-boc-amino methyl phenylboronic acid, 3-n-boc aminomethylphenylboronic acid, 3-tert-butoxycarbonylamino methyl phenylboronic acid, 3-n-boc-aminomethyl phenyl boronic acid, 3-tert-butoxycarbonylaminomethyl phenylboronic acid, 3-2-methylpropan-2-yl oxycarbonylamino methyl phenyl boronic acid, 3-aminomethyl benzeneboronic acid, n-boc protected, 3-tert-butoxycarbonyl amino methyl phenylboronic acid
PubChem CID 3684686
IUPAC Name [3-[[(2-methylpropan-2-yl)oxycarbonylamino]methyl]phenyl]boronic acid
SMILES B(C1=CC(=CC=C1)CNC(=O)OC(C)(C)C)(O)O

Specifications

Melting Point 188°C to 190°C
Quantity 250 mg
Formula Weight 251.09
Percent Purity 97%
Chemical Name or Material 3-(Boc-aminomethyl)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only