missing translation for 'onlineSavingsMsg'
Learn More

3-(BOC-amino)propyl bromide, 95%

Catalog No. p-3777550
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC397010010 1 g Glass Bottle
AC397010050 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC397010010 Supplier Thermo Scientific Chemicals Supplier No. 397010010

CAS: 83948-53-2 | C8H16BrNO2 | 238.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 83948-53-2
Molecular Formula C8H16BrNO2
Molecular Weight (g/mol) 238.13
MDL Number MFCD02683429
InChI Key IOKGWQZQCNXXLD-UHFFFAOYSA-N
Synonym 3-boc-amino propyl bromide, tert-butyl 3-bromopropylcarbamate, tert-butyl n-3-bromopropyl carbamate, 3-boc-amino propylbromide, tert-butyl 3-bromopropyl carbamate, 3-bromo-propyl-carbamic acid tert-butyl ester, n-3-bromopropyl carbamic acid tert-butyl ester, 3-tert-butoxycarbonylamino propyl bromide, n-boc-3-bromopropylamine, 1-n-boc-3-bromopropylamine
PubChem CID 4460490
IUPAC Name tert-butyl N-(3-bromopropyl)carbamate
SMILES CC(C)(C)OC(=O)NCCCBr

Specifications

Melting Point 34°C to 36.5°C
CAS Min % 94.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 238.12
Percent Purity 95%
Physical Form Low Melting Solid
Chemical Name or Material 3-(BOC-amino)propyl bromide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only