missing translation for 'onlineSavingsMsg'
Learn More

3-(Boc-amino)cyclohexanone, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6659406

 View more versions of this product

Catalog No. AAH6659406


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-(Boc-amino)cyclohexanone
C11H19NO3
11040732
VGDCXKATFLOEHF-UHFFFAOYNA-N
tert-butyl N-(3-oxocyclohexyl)carbamate
213.28
213.28
Solid
885280-38-6
MFCD03790905
3-n-boc-aminocyclohexanone, tert-butyl 3-oxocyclohexylcarbamate, 3-oxo-cyclohexyl-carbamic acid tert-butyl ester, tert-butyl 3-oxocyclohexyl carbamate, 3-boc-amino cyclohexanone, tert-butyl n-3-oxocyclohexyl carbamate, tert-butyl3-oxocyclohexylcarbamate, pubchem20066, 3-n-boc-amino cyclohexanone, 3-boc-amino cyclohexan-1-one
CC(C)(C)OC(=O)NC1CCCC(=O)C1
5 g
44228975
95%

Chemical Identifiers

885280-38-6
213.28
VGDCXKATFLOEHF-UHFFFAOYNA-N
44228975
CC(C)(C)OC(=O)NC1CCCC(=O)C1
C11H19NO3
MFCD03790905
3-n-boc-aminocyclohexanone, tert-butyl 3-oxocyclohexylcarbamate, 3-oxo-cyclohexyl-carbamic acid tert-butyl ester, tert-butyl 3-oxocyclohexyl carbamate, 3-boc-amino cyclohexanone, tert-butyl n-3-oxocyclohexyl carbamate, tert-butyl3-oxocyclohexylcarbamate, pubchem20066, 3-n-boc-amino cyclohexanone, 3-boc-amino cyclohexan-1-one
tert-butyl N-(3-oxocyclohexyl)carbamate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only