accessibility menu, dialog, popup

UserName

CAS RN 885280-38-6

IUPAC Name: tert-butyl N-(3-oxocyclohexyl)carbamate
Synonyms: tert-butyl N-(3-oxocyclohexyl)carbamate
Molecular Weight (g/mol): 213.28
Molecular Formula: C11H19NO3
InChi Key: VGDCXKATFLOEHF-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC1CCCC(=O)C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)