missing translation for 'onlineSavingsMsg'
Learn More

3-(Boc-amino)cyclohexanecarboxylic acid, 98+%

Catalog No. p-7034610
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5229903 1 g
AAH5229906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5229903 Supplier Thermo Scientific Chemicals Supplier No. H5229903
Only null left

CAS: 334932-13-7 | C12H21NO4 | 243.303 g/mol

3-(Boc-amino)cyclohexanecarboxylic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Boc-amino)cyclohexanecarboxylic acid is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water (2.1 g/L at 25°C).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 334932-13-7
Molecular Formula C12H21NO4
Molecular Weight (g/mol) 243.303
MDL Number MFCD03453252
InChI Key JSGHMGKJNZTKGF-UHFFFAOYSA-N
Synonym 3-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 3-boc-amino cyclohexanecarboxylic acid, 3-tert-butoxy carbonyl amino cyclohexane-1-carboxylic acid, 3-2-methylpropan-2-yl oxycarbonylamino cyclohexane-1-carboxylic acid, cyclohexanecarboxylic acid, 3-1,1-dimethylethoxy carbonyl amino, cis-3-boc-amino cyclohexanecarboxylic acid, trans-3-tert-butoxycarbonylamino cyclohexanecarboxylic acid, 3-tert-butoxycarbonyl amino cyclohexane-1-carboxylic acid, acmc-209fp1
PubChem CID 4420316
IUPAC Name 3-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1CCCC(C1)C(=O)O

Specifications

Solubility Information Slightly soluble in water (2.1g/L at 25°C).
Quantity 1 g
Formula Weight 243.3
Percent Purity ≥98%
Chemical Name or Material 3-(Boc-amino)cyclohexanecarboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only