accessibility menu, dialog, popup

UserName

CAS RN 334932-13-7

CH3H3CCH3OOHNOHO

CAS RN 334932-13-7

IUPAC Name: 3-{[(tert-butoxy)carbonyl]amino}cyclohexane-1-carboxylic acid
Synonyms: 3-[(tert-butoxycarbonyl)amino]cyclohexane-1-carboxylic acid
Molecular Weight (g/mol): 243.3
Molecular Formula: C12H21NO4
InChi Key: JSGHMGKJNZTKGF-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC1CCCC(C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)