missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD10/CD19 Antibody Cocktail, FITC, PE
GREENER_CHOICE

Catalog No. p-7168095
Change view
Click to view available options
Quantity:
100 Tests
50 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01670897 100 Tests
01670898 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01670897 Supplier Invitrogen™ Supplier No. MA112187

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Antibody Cocktail

This antibody is a monoclonal mixture of FITC-conjugated CD10 (clone RXB.14, isotype IgG1) and PE-conjugated CD19 (clone 1G9, isotype IgG1).

Anti-hCD10 recognizes a human common acute lymphoblastic antigen (CALLA), MW 100 kDa. The CD10 antigen is identical to human membrane-associated neutral endopeptidase (NEP;EC 3.3.24.11), also known as enkephalinase. The CD10 antigen is found on lymphocytes from patients with acute B-lymphoid leukemia. The antigen is also present on a wide variety of normal and neoplastic cell types including renal epithellum, fibroblasts, granulocytes and some lymphoma, melanoma and glioma cell lines. Identification of CD19 PE human B cells associated approximately 10% of peripheral blood lymphocytes expressing the 95,000 M.W. surface antigen.

Specifications

Antigen CD10/CD19
Applications Flow Cytometry
Classification Cocktail
Clone RXB.14, 1G9, 1G9
Conjugate FITC/PE
Formulation PBS with 0.2% carrier protein and 0.08% sodium azide
Gene MME
Gene Accession No. P08473, P15391
Gene Alias atriopeptidase; AW495831; B4; B-lymphocyte antigen CD19; B-lymphocyte surface antigen B4; CALLA; CD10; CD19; CD19 molecule; CMT2T; Common acute lymphocytic leukemia antigen; CVID3; differentiation antigen CD19; Enkephalinase; EPN; membrane metallo-endopeptidase; membrane metallo-endopeptidase (neutral endopeptidase, enkephalinase, CALLA, CD10); membrane metallo-endopeptidase variant 1; membrane metallo-endopeptidase variant 2; MME; NEP; Neprilysin; neprilysin-390; neprilysin-411; neutral endopeptidase 24.11; SFE; Skin fibroblast elastase; T-cell surface antigen Leu-12
Gene Symbols CD19, MME
Host Species Mouse
Immunogen White blood cells.
Purification Method Protein A/G
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4311, 930
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less