missing translation for 'onlineSavingsMsg'
Learn More

3-(Boc-amino)-3-phenylpropionic acid, 97%

Catalog No. AAL1980203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1980203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1980203 Supplier Thermo Scientific Chemicals Supplier No. L1980203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 14676-01-8 | C14H19NO4 | 265.309 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14676-01-8
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.309
MDL Number MFCD02090695
InChI Key JTNQFJPZRTURSI-UHFFFAOYSA-N
Synonym 3-boc-amino-3-phenylpropionic acid, 3-n-boc-3-phenylpropionic acid, boc-dl-beta-phe-oh, 3-tert-butoxycarbonyl amino-3-phenylpropanoic acid, 3-boc-amino-3-phenyl-propionic acid, 3-tert-butoxycarbonylamino-3-phenylpropanoic acid, 3-tert-butoxy carbonyl amino-3-phenylpropanoic acid, boc-d-, a-phe-oh, pubchem16181
PubChem CID 2756815
IUPAC Name 3-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)O)C1=CC=CC=C1

Specifications

Melting Point 142°C to 144°C (decomposition)
Quantity 1 g
Beilstein 2867023
Formula Weight 265.31
Percent Purity 97%
Chemical Name or Material 3-(Boc-amino)-3-phenylpropionic acid
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only