accessibility menu, dialog, popup

UserName

CAS RN 14676-01-8

IUPAC Name: (3R)-3-(benzylazaniumyl)butanoate
Synonyms: (3R)-3-(benzylammonio)butanoate
Molecular Weight (g/mol): 193.25
Molecular Formula: C11H15NO2
InChi Key: QIXZNMWEYBPTEQ-SECBINFHSA-N
SMILES: C[C@H](CC([O-])=O)[NH2+]CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results