missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyloxypyrrolidine, 96%

Catalog No. AAH50769MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH50769MD 250 mg
AAH5076903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH50769MD Supplier Thermo Scientific Chemicals Supplier No. H50769MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 420137-14-0 | C11H15NO | 177.247 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 420137-14-0
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.247
MDL Number MFCD08060975
InChI Key CWBMYKUPMLRKQK-UHFFFAOYSA-N
Synonym 3-benzyloxy pyrrolidine, 3-benzyloxypyrrolidine, 3-benzyloxy-pyrrolidine, 3-phenylmethoxy pyrrolidine, benzyl pyrrolidin-3-yl ether, pyrrolidin-3-yl oxy methyl benzene
PubChem CID 14497353
IUPAC Name 3-phenylmethoxypyrrolidine
SMILES C1CNCC1OCC2=CC=CC=C2

Specifications

Quantity 250 mg
Formula Weight 177.25
Percent Purity 96%
Chemical Name or Material 3-Benzyloxypyrrolidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only