missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyloxyphenylhydrazine hydrochloride, 98%

Catalog No. AC394020010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC394020010 1 g Glass bottle
AC394020050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC394020010 Supplier Thermo Scientific Chemicals Supplier No. 394020010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 59146-68-8 | C13H15ClN2O | 250.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 59146-68-8
Molecular Formula C13H15ClN2O
Molecular Weight (g/mol) 250.73
MDL Number MFCD02258768
InChI Key MMMGCMOISVZVKZ-UHFFFAOYSA-N
Synonym 3-benzyloxyphenylhydrazine hydrochloride, 3-benzyloxy phenyl hydrazine hydrochloride, 3-benzyloxy-phenyl-hydrazine hydrochloride, 3-benzyloxyphenyl hydrazine hydrochloride, 3-benzyloxyphenylhydrazine hcl, benzyloxy-phenyl-hydrazine hydrochloride, 3-phenylmethoxyphenyl diazane hydrochloride, n'-3-benzyloxy-phenyl-hydrazinium, chloride
PubChem CID 12891839
IUPAC Name (3-phenylmethoxyphenyl)hydrazine;hydrochloride
SMILES Cl.NNC1=CC(OCC2=CC=CC=C2)=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 155.0°C to 157.0°C
Color Violet
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH2OC6H4NHNH2·HCl
Quantity 1 g
Formula Weight 250.73
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-Benzyloxyphenylhydrazine hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only