missing translation for 'onlineSavingsMsg'
Learn More

3-(Benzyloxymethyl)cyclobutanone, 97%, Thermo Scientific Chemicals

Catalog No. AAH6215903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6215903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6215903 Supplier Thermo Scientific Chemicals Supplier No. H6215903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 172324-67-3
Molecular Formula C12H14O2
Molecular Weight (g/mol) 190.24
MDL Number MFCD09055138
InChI Key FHBBBGYOVFMVIB-UHFFFAOYSA-N
PubChem CID 11819850
IUPAC Name 3-(phenylmethoxymethyl)cyclobutan-1-one
SMILES O=C1CC(COCC2=CC=CC=C2)C1

Specifications

Quantity 1 g
Synonym 3-benzyloxy methyl cyclobutanone, 3-benzyloxymethyl cyclobutanone, 3-benzyloxy methyl cyclobutan-1-one, cyclobutanone, 3-phenylmethoxy methyl, 3-benzyloxymethyl-cyclobutanone, 3-benzyloxymethyl cyclobutan-1-one, cyclobutanone,3-phenylmethoxy methyl
Formula Weight 190.23
Percent Purity 97%
Chemical Name or Material 3-(Benzyloxymethyl)cyclobutanone
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only