missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyloxybenzoic acid, 98%

Catalog No. AAH3288203
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3288203 1 g
AAH3288206 5 g
AAH3288214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH3288203 Supplier Thermo Scientific Chemicals Supplier No. H3288203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 69026-14-8 | C14H11O3 | 227.24 g/mol

3-Benzyloxybenzoic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Benzyloxybenzoic acid is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 69026-14-8
Molecular Formula C14H11O3
Molecular Weight (g/mol) 227.24
MDL Number MFCD00527901
InChI Key CISXCTKEQYOZAM-UHFFFAOYSA-M
Synonym 3-benzyloxy benzoic acid, 3-benzyloxybenzoic acid, benzoic acid, 3-phenylmethoxy, 3-benzyloxy-benzoic acid, 3-phenylmethoxy benzoic acid, enamine_004835, acmc-1b5k3
PubChem CID 309226
IUPAC Name 3-phenylmethoxybenzoic acid
SMILES [O-]C(=O)C1=CC=CC(OCC2=CC=CC=C2)=C1

Specifications

Melting Point 133°C to 135°C
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 228.25
Percent Purity 98%
Chemical Name or Material 3-Benzyloxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only