missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyloxybenzaldehyde 98.0+%, TCI America™

Catalog No. B240025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B240025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B240025G Supplier TCI America Supplier No. B240025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1700-37-4
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.248
MDL Number MFCD00003367
InChI Key JAICGBJIBWDEIZ-UHFFFAOYSA-N
Synonym 3-benzyloxybenzaldehyde, 3-benzyloxy benzaldehyde, benzaldehyde, 3-phenylmethoxy, m-benzyloxybenzaldehyde, benzaldehyde, m-benzyloxy, 3-phenylmethoxy benzaldehyde, 3-benzyloxy-benzaldehyde, unii-k1s6ob2gnn, k1s6ob2gnn
PubChem CID 74342
IUPAC Name 3-phenylmethoxybenzaldehyde
SMILES C1=CC=C(C=C1)COC2=CC=CC(=C2)C=O

Specifications

Melting Point 57°C
Color White-Yellow
Quantity 25 g
Formula Weight 212.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Benzyloxybenzaldehyde