accessibility menu, dialog, popup

UserName

CAS RN 1700-37-4

IUPAC Name: 3-(benzyloxy)benzaldehyde
Synonyms: 3-(benzyloxy)benzaldehyde
Molecular Weight (g/mol): 212.25
Molecular Formula: C14H12O2
InChi Key: JAICGBJIBWDEIZ-UHFFFAOYSA-N
SMILES: O=CC1=CC=CC(OCC2=CC=CC=C2)=C1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)