missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyloxybenzaldehyde, 98%

Catalog No. AAB2290906
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2290906 5 g
AAB2290914 25 g
AAB2290922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2290906 Supplier Thermo Scientific Chemicals Supplier No. B2290906

CAS: 1700-37-4 | C14H12O2 | 212.248 g/mol

3-Benzyloxybenzaldehyde is used as a pharmaceutical intermediate. It activates the adenyl cyclase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Benzyloxybenzaldehyde is used as a pharmaceutical intermediate. It activates the adenyl cyclase.

Solubility
Soluble in water 38.87 mg/L @ 25°C.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 1700-37-4
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.248
MDL Number MFCD00003367
InChI Key JAICGBJIBWDEIZ-UHFFFAOYSA-N
Synonym 3-benzyloxybenzaldehyde, 3-benzyloxy benzaldehyde, benzaldehyde, 3-phenylmethoxy, m-benzyloxybenzaldehyde, benzaldehyde, m-benzyloxy, 3-phenylmethoxy benzaldehyde, 3-benzyloxy-benzaldehyde, unii-k1s6ob2gnn, k1s6ob2gnn
PubChem CID 74342
IUPAC Name 3-phenylmethoxybenzaldehyde
SMILES C1=CC=C(C=C1)COC2=CC=CC(=C2)C=O

Specifications

Melting Point 54°C to 58°C
Boiling Point 180°C (0.08 mmHg)
Quantity 5 g
Beilstein 1956589
Sensitivity Air Sensitive
Solubility Information Soluble in water 38.87mg/L @ 25°C.
Formula Weight 212.25
Percent Purity 98%
Chemical Name or Material 3-Benzyloxybenzaldehyde
EINECSNumber 216-932-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only