missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyloxyaniline 98.0+%, TCI America™

Catalog No. p-7123404
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
B41975G 5 g
B419725G 25 g
2 options

Catalog No. B41975G

Supplier: TCI America B41975G

Only null left

Chemical Identifiers

CAS 1484-26-0
Molecular Formula C13H13NO
Molecular Weight (g/mol) 199.253
MDL Number MFCD00007784
InChI Key IGPFOKFDBICQMC-UHFFFAOYSA-N
Synonym 3-benzyloxyaniline, 3-benzyloxy aniline, benzenamine, 3-phenylmethoxy, unii-iab1vda972, 3-benzyloxy-phenylamine, iab1vda972, 3-phenylmethyl oxy aniline, 3-phenylmethoxy phenylamine, 3-benzyloxy benzenamine, 3-benyloxyaniline
PubChem CID 92892
IUPAC Name 3-phenylmethoxyaniline
SMILES C1=CC=C(C=C1)COC2=CC=CC(=C2)N

Specifications

Melting Point 66°C
Color White-Yellow
Quantity 5 g
Formula Weight 199.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Benzyloxyaniline