missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyloxyaniline, 98%

Catalog No. p-7052766
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Catalog No. Quantity
AAH5570803 1 g
AAH5570806 5 g
AAH5570814 25 g
3 options

Catalog No. AAH5570803

Supplier: Thermo Scientific Chemicals H5570803

Only null left

CAS: 1484-26-0 | C13H13NO | 199.253 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1484-26-0
Molecular Formula C13H13NO
Molecular Weight (g/mol) 199.253
MDL Number MFCD00007784
InChI Key IGPFOKFDBICQMC-UHFFFAOYSA-N
Synonym 3-benzyloxyaniline, 3-benzyloxy aniline, benzenamine, 3-phenylmethoxy, unii-iab1vda972, 3-benzyloxy-phenylamine, iab1vda972, 3-phenylmethyl oxy aniline, 3-phenylmethoxy phenylamine, 3-benzyloxy benzenamine, 3-benyloxyaniline
PubChem CID 92892
IUPAC Name 3-phenylmethoxyaniline
SMILES C1=CC=C(C=C1)COC2=CC=CC(=C2)N

Specifications

Melting Point 63°C to 67°C
Color Brown
Quantity 1 g
Formula Weight 199.25
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-Benzyloxyaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 216-056-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only