Learn More
3-Azetidinemethanol hydrochloride, 95%
CAS: 928038-44-2 | C4H10ClNO | 123.58 g/mol
$165.77 - $303.63
Chemical Identifiers
| CAS | 928038-44-2 |
|---|---|
| Molecular Formula | C4H10ClNO |
| Molecular Weight (g/mol) | 123.58 |
| MDL Number | MFCD08703352 |
| InChI Key | AQUVQGSNKVDBBF-UHFFFAOYSA-N |
| Synonym | azetidin-3-ylmethanol hydrochloride, 3-azetidinemethanol hydrochloride, azetidine-3-yl methanol hcl, 3-hydroxymethyl-azetidine hcl, azetidin-3-yl methanol hydrochloride, 3-hydroxymethyl azetidine hydrochloride, 3-azetidinemethanol, hydrochloride 1:1, azetidine-3-yl methanol hydrochloride, pubchem19094, 3-hydroxymethyl azetidine 3 |
| PubChem CID | 24801400 |
| IUPAC Name | azetidin-3-ylmethanol;hydrochloride |
| SMILES | C1C(CN1)CO.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH57801MD
|
Thermo Scientific Chemicals
H57801MD |
250 mg |
Each for $165.77
|
|
|||||
|
AAH57801ME
|
Thermo Scientific Chemicals
H57801ME |
500 mg |
Each for $303.63
|
|
|||||
Description
3-Azetidinemethanol Hydrochloride is used in the synthesis of human rhinovirus inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Azetidinemethanol Hydrochloride is used in the synthesis of human rhinovirus inhibitors.
Notes
Hygroscopic. Store at room temperature.
Chemical Identifiers
| 928038-44-2 | |
| 123.58 | |
| AQUVQGSNKVDBBF-UHFFFAOYSA-N | |
| 24801400 | |
| C1C(CN1)CO.Cl |
| C4H10ClNO | |
| MFCD08703352 | |
| azetidin-3-ylmethanol hydrochloride, 3-azetidinemethanol hydrochloride, azetidine-3-yl methanol hcl, 3-hydroxymethyl-azetidine hcl, azetidin-3-yl methanol hydrochloride, 3-hydroxymethyl azetidine hydrochloride, 3-azetidinemethanol, hydrochloride 1:1, azetidine-3-yl methanol hydrochloride, pubchem19094, 3-hydroxymethyl azetidine 3 | |
| azetidin-3-ylmethanol;hydrochloride |
Specifications
| 928038-44-2 | |
| MFCD08703352 | |
| Hygroscopic | |
| AQUVQGSNKVDBBF-UHFFFAOYSA-N | |
| azetidin-3-ylmethanol;hydrochloride | |
| 24801400 | |
| 95% |
| C4H10ClNO | |
| 250 mg | |
| azetidin-3-ylmethanol hydrochloride, 3-azetidinemethanol hydrochloride, azetidine-3-yl methanol hcl, 3-hydroxymethyl-azetidine hcl, azetidin-3-yl methanol hydrochloride, 3-hydroxymethyl azetidine hydrochloride, 3-azetidinemethanol, hydrochloride 1:1, azetidine-3-yl methanol hydrochloride, pubchem19094, 3-hydroxymethyl azetidine 3 | |
| C1C(CN1)CO.Cl | |
| 123.58 | |
| 123.58 | |
| 3-Azetidinemethanol hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only