Learn More
3-Azetidinemethanol hydrochloride, 95%
CAS: 928038-44-2 | C4H10ClNO | 123.58 g/mol
Supplier: Thermo Scientific Chemicals H57801MD
Description
3-Azetidinemethanol Hydrochloride is used in the synthesis of human rhinovirus inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Azetidinemethanol Hydrochloride is used in the synthesis of human rhinovirus inhibitors.
Notes
Hygroscopic. Store at room temperature.
Specifications
| 3-Azetidinemethanol hydrochloride | |
| C4H10ClNO | |
| 250 mg | |
| azetidin-3-ylmethanol hydrochloride, 3-azetidinemethanol hydrochloride, azetidine-3-yl methanol hcl, 3-hydroxymethyl-azetidine hcl, azetidin-3-yl methanol hydrochloride, 3-hydroxymethyl azetidine hydrochloride, 3-azetidinemethanol, hydrochloride 1:1, azetidine-3-yl methanol hydrochloride, pubchem19094, 3-hydroxymethyl azetidine 3 | |
| C1C(CN1)CO.Cl | |
| 123.58 | |
| 123.58 |
| 928038-44-2 | |
| MFCD08703352 | |
| Hygroscopic | |
| AQUVQGSNKVDBBF-UHFFFAOYSA-N | |
| azetidin-3-ylmethanol;hydrochloride | |
| 24801400 | |
| 95% |
Chemical Identifiers
| 928038-44-2 | |
| 123.58 | |
| AQUVQGSNKVDBBF-UHFFFAOYSA-N | |
| 24801400 | |
| C1C(CN1)CO.Cl |
| C4H10ClNO | |
| MFCD08703352 | |
| azetidin-3-ylmethanol hydrochloride, 3-azetidinemethanol hydrochloride, azetidine-3-yl methanol hcl, 3-hydroxymethyl-azetidine hcl, azetidin-3-yl methanol hydrochloride, 3-hydroxymethyl azetidine hydrochloride, 3-azetidinemethanol, hydrochloride 1:1, azetidine-3-yl methanol hydrochloride, pubchem19094, 3-hydroxymethyl azetidine 3 | |
| azetidin-3-ylmethanol;hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only