missing translation for 'onlineSavingsMsg'
Learn More

3-Anilino-1-propanol 98.0+%, TCI America™

Catalog No. A22485G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A22485G 5 g
A224825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A22485G Supplier TCI America Supplier No. A22485G
Only null left

Chemical Identifiers

CAS 31121-11-6
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
MDL Number MFCD15206074
InChI Key GZYBSURBYCLNSM-UHFFFAOYSA-N
Synonym N-(3-Hydroxypropyl)aniline
PubChem CID 410467
IUPAC Name 3-(phenylamino)propan-1-ol
SMILES OCCCNC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 140°C
Quantity 5 g
Formula Weight 151.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Anilino-1-propanol
TSCA No
Recommended Storage Refrigerator