accessibility menu, dialog, popup

UserName

CAS RN 31121-11-6

HNOH

CAS RN 31121-11-6

IUPAC Name: 3-(phenylamino)propan-1-ol
Synonyms: 1-propanol, 3-(phenylamino)-
Molecular Weight (g/mol): 151.21
Molecular Formula: C9H13NO
InChi Key: GZYBSURBYCLNSM-UHFFFAOYSA-N
SMILES: OCCCNC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results