missing translation for 'onlineSavingsMsg'
Learn More

3-Aminothiobenzamide, 97%

Catalog No. AAH5180603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5180603 1 g
AAH5180606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5180603 Supplier Thermo Scientific Chemicals Supplier No. H5180603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 78950-36-4 | C7H8N2S | 152.215 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 78950-36-4
Molecular Formula C7H8N2S
Molecular Weight (g/mol) 152.215
MDL Number MFCD04973325
InChI Key ZKWTUTBIHCNCKU-UHFFFAOYSA-N
Synonym 3-aminothiobenzamide, 3-aminobenzothioamide, 3-amino-thiobenzamide, 3-aminobenzene-1-carbothioamide, 3-aminobenzthioamide, 3-azanylbenzenecarbothioamide, #, benzenecarbothioamide,3-amino, 3-amino-benzenecarbothioamide, amino 3-aminophenyl methane-1-thione
PubChem CID 2060897
IUPAC Name 3-aminobenzenecarbothioamide
SMILES C1=CC(=CC(=C1)N)C(=S)N

Specifications

Melting Point 127°C to 131°C
Quantity 1 g
Sensitivity Air and moisture sensitive
Formula Weight 152.21
Percent Purity 97%
Chemical Name or Material 3-Aminothiobenzamide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only