missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Aminosalicylic Acid 98.0+%, TCI America™
$197.59 - $656.14
Chemical Identifiers
| CAS | 570-23-0 |
|---|---|
| Molecular Formula | C7H7NO3 |
| Molecular Weight (g/mol) | 153.137 |
| MDL Number | MFCD00010299 |
| InChI Key | IQGMRVWUTCYCST-UHFFFAOYSA-N |
| Synonym | 3-aminosalicylic acid, 3-amino salicylic acid, benzoic acid, 3-amino-2-hydroxy, 3-amino-2-hydroxy-benzoic acid, 2-amino-6-carboxyphenol, salicylic acid, 3-amino, 3-carboxy-2-hydroxyaniline, 3 or 5-aminosalicylic acid, benzoic acid, amino-2-hydroxy |
| PubChem CID | 68443 |
| IUPAC Name | 3-amino-2-hydroxybenzoic acid |
| SMILES | C1=CC(=C(C(=C1)N)O)C(=O)O |
Chemical Identifiers
| 570-23-0 | |
| 153.137 | |
| IQGMRVWUTCYCST-UHFFFAOYSA-N | |
| 68443 | |
| C1=CC(=C(C(=C1)N)O)C(=O)O |
| C7H7NO3 | |
| MFCD00010299 | |
| 3-aminosalicylic acid, 3-amino salicylic acid, benzoic acid, 3-amino-2-hydroxy, 3-amino-2-hydroxy-benzoic acid, 2-amino-6-carboxyphenol, salicylic acid, 3-amino, 3-carboxy-2-hydroxyaniline, 3 or 5-aminosalicylic acid, benzoic acid, amino-2-hydroxy | |
| 3-amino-2-hydroxybenzoic acid |
Specifications
| 570-23-0 | |
| C7H7NO3 | |
| 1 g | |
| IQGMRVWUTCYCST-UHFFFAOYSA-N | |
| 3-amino-2-hydroxybenzoic acid | |
| 68443 | |
| ≥98.0% (HPLC) | |
| 3-Aminosalicylic Acid |
| 221°C | |
| MFCD00010299 | |
| 3-aminosalicylic acid, 3-amino salicylic acid, benzoic acid, 3-amino-2-hydroxy, 3-amino-2-hydroxy-benzoic acid, 2-amino-6-carboxyphenol, salicylic acid, 3-amino, 3-carboxy-2-hydroxyaniline, 3 or 5-aminosalicylic acid, benzoic acid, amino-2-hydroxy | |
| C1=CC(=C(C(=C1)N)O)C(=O)O | |
| 153.137 | |
| 153.14 | |
| Crystalline Powder |
Safety and Handling
TSCA : No