missing translation for 'onlineSavingsMsg'
Learn More

3-Aminosalicylic acid, 97%

CAS: 570-23-0 | C7H7NO3 | 153.137 g/mol

Supplier:  Thermo Scientific Chemicals B2247802

 View more versions of this product

Catalog No. AAB2247802


Only null left
Explore available promotions

Description

Description

3-Aminosalicylic acid acts as an oxygen, nitrogen donor ligand and forms complexes with silver.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminosalicylic acid acts as an oxygen, nitrogen donor ligand and forms complexes with silver.

Solubility
Soluble in potassium hydroxide. Slightly soluble in water.

Notes
Air and light sensitive. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications

Specifications

3-Aminosalicylic acid
∼240°C (decomposition)
MFCD00010299
2090427
Soluble in potassium hydroxide. Slightly soluble in water.
C1=CC(=C(C(=C1)N)O)C(=O)O
153.137
153.14
570-23-0
C7H7NO3
0.5 g
3-aminosalicylic acid, 3-amino salicylic acid, benzoic acid, 3-amino-2-hydroxy, 3-amino-2-hydroxy-benzoic acid, 2-amino-6-carboxyphenol, salicylic acid, 3-amino, 3-carboxy-2-hydroxyaniline, 3 or 5-aminosalicylic acid, benzoic acid, amino-2-hydroxy
IQGMRVWUTCYCST-UHFFFAOYSA-N
3-amino-2-hydroxybenzoic acid
68443
97%

Chemical Identifiers

570-23-0
153.137
IQGMRVWUTCYCST-UHFFFAOYSA-N
68443
C1=CC(=C(C(=C1)N)O)C(=O)O
C7H7NO3
MFCD00010299
3-aminosalicylic acid, 3-amino salicylic acid, benzoic acid, 3-amino-2-hydroxy, 3-amino-2-hydroxy-benzoic acid, 2-amino-6-carboxyphenol, salicylic acid, 3-amino, 3-carboxy-2-hydroxyaniline, 3 or 5-aminosalicylic acid, benzoic acid, amino-2-hydroxy
3-amino-2-hydroxybenzoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 209-328-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only