missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Aminopyrrolidine 99.0+%, TCI America™
$201.32 - $602.10
Chemical Identifiers
| CAS | 79286-79-6 |
|---|---|
| Molecular Formula | C4H10N2 |
| Molecular Weight (g/mol) | 86.138 |
| MDL Number | MFCD00059018 |
| InChI Key | NGXSWUFDCSEIOO-UHFFFAOYSA-N |
| PubChem CID | 164401 |
| IUPAC Name | pyrrolidin-3-amine |
| SMILES | C1CNCC1N |
Chemical Identifiers
| 79286-79-6 | |
| 86.138 | |
| NGXSWUFDCSEIOO-UHFFFAOYSA-N | |
| pyrrolidin-3-amine |
| C4H10N2 | |
| MFCD00059018 | |
| 164401 | |
| C1CNCC1N |
Specifications
| 79286-79-6 | |
| C4H10N2 | |
| 5 g | |
| NGXSWUFDCSEIOO-UHFFFAOYSA-N | |
| pyrrolidin-3-amine | |
| 164401 | |
| ≥99.0% (GC,T) | |
| 3-Aminopyrrolidine |
| Yellow | |
| MFCD00059018 | |
| 2735 | |
| C1CNCC1N | |
| 86.138 | |
| 86.14 | |
| Liquid |
Safety and Handling
TSCA : No