CAS RN 79286-79-6
CAS RN 79286-79-6
IUPAC Name:
pyrrolidin-3-amine
Synonyms:
pyrrolidin-3-amine
Molecular Weight (g/mol):
86.14
Molecular Formula:
C4H10N2
InChi Key:
NGXSWUFDCSEIOO-UHFFFAOYNA-N
SMILES:
NC1CCNC1
1
–
1
of
1
results
1
3-Aminopyrrolidine 99.0+%, TCI America™
CAS: 79286-79-6 Molecular Formula: C4H10N2 Molecular Weight (g/mol): 86.138 MDL Number: MFCD00059018 InChI Key: NGXSWUFDCSEIOO-UHFFFAOYSA-N PubChem CID: 164401 IUPAC Name: pyrrolidin-3-amine SMILES: C1CNCC1N