missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Aminopyrrolidine Dihydrochloride 98.0+%, TCI America™
$126.76 - $410.10
Chemical Identifiers
| CAS | 103831-11-4 |
|---|---|
| Molecular Formula | C4H12Cl2N2 |
| Molecular Weight (g/mol) | 159.054 |
| MDL Number | MFCD00060176 |
| InChI Key | NJPNCMOUEXEGBL-UHFFFAOYSA-N |
| Synonym | 3-aminopyrrolidine dihydrochloride, pyrrolidin-3-amine dihydrochloride, 3-pyrrolidinamine, dihydrochloride, 3-aminopyrrolidine 2hcl, 3-pyrrolidinamine dihydrochloride, 3-amino-pyrrolidine dihydrochloride, ?-3-aminopyrrolidine dihydrochloride, pyrrolidine-3-ylamine, chloride, chloride, pyrrolidin-3-amine-hydrogen chloride 1/2 |
| PubChem CID | 16212596 |
| IUPAC Name | pyrrolidin-3-amine;dihydrochloride |
| SMILES | C1CNCC1N.Cl.Cl |
Chemical Identifiers
| 103831-11-4 | |
| 159.054 | |
| NJPNCMOUEXEGBL-UHFFFAOYSA-N | |
| 16212596 | |
| C1CNCC1N.Cl.Cl |
| C4H12Cl2N2 | |
| MFCD00060176 | |
| 3-aminopyrrolidine dihydrochloride, pyrrolidin-3-amine dihydrochloride, 3-pyrrolidinamine, dihydrochloride, 3-aminopyrrolidine 2hcl, 3-pyrrolidinamine dihydrochloride, 3-amino-pyrrolidine dihydrochloride, ?-3-aminopyrrolidine dihydrochloride, pyrrolidine-3-ylamine, chloride, chloride, pyrrolidin-3-amine-hydrogen chloride 1/2 | |
| pyrrolidin-3-amine;dihydrochloride |
Specifications
| 103831-11-4 | |
| MFCD00060176 | |
| 3-aminopyrrolidine dihydrochloride, pyrrolidin-3-amine dihydrochloride, 3-pyrrolidinamine, dihydrochloride, 3-aminopyrrolidine 2hcl, 3-pyrrolidinamine dihydrochloride, 3-amino-pyrrolidine dihydrochloride, ?-3-aminopyrrolidine dihydrochloride, pyrrolidine-3-ylamine, chloride, chloride, pyrrolidin-3-amine-hydrogen chloride 1/2 | |
| C1CNCC1N.Cl.Cl | |
| 159.054 | |
| 159.05 | |
| Crystalline Powder |
| C4H12Cl2N2 | |
| 5 g | |
| NJPNCMOUEXEGBL-UHFFFAOYSA-N | |
| pyrrolidin-3-amine;dihydrochloride | |
| 16212596 | |
| ≥98.0% (HPLC,N) | |
| 3-Aminopyrrolidine Dihydrochloride |
Safety and Handling
TSCA : No