missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopyridine 99.0+%, TCI America™

Catalog No. A041325G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A041325G 25 g
A0413100G 100 g
A0413500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A041325G Supplier TCI America Supplier No. A041325G

Chemical Identifiers

CAS 462-08-8
Molecular Formula C5H6N2
Molecular Weight (g/mol) 94.12
MDL Number MFCD00006400
InChI Key CUYKNJBYIJFRCU-UHFFFAOYSA-N
Synonym 3-aminopyridine, 3-pyridinamine, 3-pyridylamine, pyridin-3-ylamine, m-aminopyridine, amino-3 pyridine, pyridine, 3-amino, 3-amino pyridine, beta-aminopyridine, amino-3-pyridine
PubChem CID 10009
IUPAC Name pyridin-3-amine
SMILES NC1=CC=CN=C1

Specifications

Melting Point 62°C
pH 10 (100g/L H2O soln. 20°C)
Boiling Point 248°C
Quantity 25 g
UN Number 2671
Formula Weight 94.12
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminopyridine
EINECSNumber (5)-0725
RTECSNumber US1650000
TSCA Yes