accessibility menu, dialog, popup

UserName

NH2N

CAS RN 462-08-8

IUPAC Name: pyridin-3-amine
Synonyms: 3-aminopyridine
Molecular Weight (g/mol): 94.12
Molecular Formula: C5H6N2
InChi Key: CUYKNJBYIJFRCU-UHFFFAOYSA-N
SMILES: NC1=CC=CN=C1
Boiling Point 
  • (3)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)