missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopyridine, 99%

Catalog No. AC104560250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC104560250 25 g Glass bottle
AC104561000 100 g Glass bottle
AC104565000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC104560250 Supplier Thermo Scientific Chemicals Supplier No. 104560250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 462-08-8 | C5H6N2 | 94.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 462-08-8
Molecular Formula C5H6N2
Molecular Weight (g/mol) 94.12
MDL Number MFCD00006400
InChI Key CUYKNJBYIJFRCU-UHFFFAOYSA-N
Synonym 3-aminopyridine, 3-pyridinamine, 3-pyridylamine, pyridin-3-ylamine, m-aminopyridine, amino-3 pyridine, pyridine, 3-amino, 3-amino pyridine, beta-aminopyridine, amino-3-pyridine
PubChem CID 10009
IUPAC Name pyridin-3-amine
SMILES NC1=CC=CN=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 60.0°C to 63.0°C
Color Colorless to Yellow
Boiling Point 248.0°C
Flash Point 124°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (C5H4N)NH2
Quantity 25 g
Beilstein 22, 431
Merck Index 15, 469
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, benzene and ether, insoluble in petroleum ether
Formula Weight 94.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Aminopyridine, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if swallowed.
Toxic in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously wit
EINECSNumber 207-322-2
RTECSNumber US1650000
TSCA TSCA

RUO – Research Use Only